1,2,4-Trichlorobenzene ≥99%, CHROMASOLV™ for HPLC, Riedel-de Haën™
:120-82-1
:
:C₆H₃Cl₃
:MFCD00000547
:181.45 g/mol
:13,09704
:213.5 °C
:17 °C
:1.454 g/cm³ (20 °C)
:105 °C
:Ambient
:204-428-0
:2321
:6.1,III
:GHS07
:GHS09
- Environmentally Preferable:
- Appearance of the substance:complying
- Identity (IR):complying
- Assay (GC):Min. 99.0 %
- Water (Karl Fischer):Max. 0.02 %
- Absorbance at 400 nm:Max. 0.01
- Absorbance at 375 nm:Max. 0.03
- Absorbance at 350 nm:Max. 0.1
- Absorbance at 310 nm:Max. 0.50
- Absorbance at 308 nm:Max. 1.00
- Explosion Limit:6.6 %, 150 °F
- Refractive Index:n20/D 1.571 (lit.)
Frequently Bought Together