Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromophenetole
3-Bromophenetole
3-Bromophenetole
MOLEM70318195
:
Molekula
:
3-Bromophenetole
MOLEM70318195
:
Molekula
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
201.06 g/mol
:
220-186-9
:
2655-84-7
:
C₈H₉BrO
:
MFCD00274287