4-Bromophenyl isothiocyanate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1985-12-2
:
Formula:C₇H₄BrNS
MDL Number:MFCD00004808
:214.09 g/mol
Boiling Pt:144…145 °C (5 mmHg)
Melting Pt:56…60 °C
Flash Pt:>110 °C (230 °F)
Storage Temperature:Ambient
ADR:8,III
:GHS06
:GHS05
Related Information
Frequently Bought Together