Orders LinkContactUsLinkComponent
Screw cap, without dropper
Screw cap, without dropper
Screw cap, GL 18, White, PP/copolymer resin
Catalogus # MUEL14.016.19
Leverancier:   MUELLER KREMPEL
Screw cap, without dropper
Catalogus # MUEL14.016.19
Leverancier:  MUELLER KREMPEL
Screw cap, without dropper
Screw cap, without dropper
Screw cap, GL 18, White, PP/copolymer resin
Price
28,80 €
Log in voor beschikbaarheid
1 * 100 ST
Product Details & Documents
Self-sealing caps without drip insert, suitable for steam sterilization.
Specificaties
  • Thread:
    GL 18
  • Colour:
    White
  • Description:
    Screw cap
  • Material:
    PP/copolymer resin