Volumetric flasks, class A, trapezoidal shape, BLAUBRAND®
Supplier: Brand
Certificates
About this item
Borosilicate glass 3.3, with PP stopper.
- Calibrated to contain (TC, In)
- Marks and inscriptions in high contrast blue enamel
- DE-M marking
- Total order from one batch, batch certificate included
*Wide neck.
Certifications: DIN EN ISO 1042
Frequently Bought Together