Order Entry
Belgium
ContactUsLinkComponent
 
Irritant
4-Methoxyphenylboronic acid
Catalog # MOLE57628191-1G
Supplier:  Molekula
CAS Number:  
Irritant
4-Methoxyphenylboronic acid
Catalog # MOLE57628191-1G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :151.96 g/mol
Melting Pt:204…207 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:5720-07-0
Formula:C₇H₉BO₃
MDL Number:MFCD00039139