Order Entry
Belgium
ContactUsLinkComponent
Phenylboronic acid
Irritant
Phenylboronic acid
Catalog # MOLE11859927-25G
Supplier:  Molekula
CAS Number:  
Irritant
Phenylboronic acid
Catalog # MOLE11859927-25G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :121.93 g/mol
Merck Index:13,01068
Melting Pt:217…220 °C
EINECS:202-701-9
 :GHS07
Storage Temperature:Ambient
CAS Number:98-80-6
Formula:C₆H₇BO₂
MDL Number:MFCD00002103