Phenylboronic acid 98+% may contain varying amounts of anhydride
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:98-80-6
:
Formula:C₆H₇BO₂
MDL Number:MFCD00002103
:121.93 g/mol
Merck Index:13,01068
Melting Pt:217…222 °C
EINECS:202-701-9
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:217 to 222 °C
- HPLC:≥98.0 %
Specification Test Results
Frequently Bought Together