2-Methylindoline
Glass bottle for solids, 100 mg
Catalog # PROOMM0477.08
Supplier: LGC Standards PROMOCHEM
Jump to:
Product Details & Documents
- CAS Number:6872-06-6
- Formula:C₉H₁₁N
- MDL Number:MFCD00005706
- Molecular Weight:133.19 g/mol
- Boiling Pt:228 °C (1013 hPa)
- Density:1,0236 g/cm³ (15 °C)
- EINECS:229-971-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



