2-Methylindoline
Glass bottle for solids, 100 g
Catalog # MOLE13901024-100G
Marketsource
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:6872-06-6
- Formula:C₉H₁₁N
- MDL Number:MFCD00005706
- Molecular Weight:133.19 g/mol
- Boiling Pt:228 °C (1013 hPa)
- Density:1,0236 g/cm³ (15 °C)
- EINECS:229-971-0
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



