2-Methylindoline 99+%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:6872-06-6
- Formula:C₉H₁₁N
- MDL Number:MFCD00005706
- Molecular Weight:133.19 g/mol
- Boiling Pt:227 °C (1013mbar)
- Melting Pt:–51 °C
- Density:1.02 g/cm³
- Flash Pt:103 °C
- EINECS:229-971-0
2 Options Available Below
Product Details & Documents
- CAS Number:6872-06-6
- Formula:C₉H₁₁N
- MDL Number:MFCD00005706
- Molecular Weight:133.19 g/mol
- Boiling Pt:227 °C (1013mbar)
- Melting Pt:–51 °C
- Density:1.02 g/cm³
- Flash Pt:103 °C
- EINECS:229-971-0
Documents
Specifications for Product Family
Specification Test Results
- Appearance:Clear yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5680 to 1.5700 (20°C, 589 nm)
- Water:≤0.5 % (K.F.)
- GC:≥99.0 %



