Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cycloheximide ≥90.0%

Cycloheximide ≥90.0%

Glass bottle, 10 g
N° de catalogue 441894C
Health
Toxic
Environment
Fournisseur:   VWR Chemicals
Cycloheximide ≥90.0%
N° de catalogue 441894C
Fournisseur:  VWR Chemicals
Numéro CAS:  66-81-9
Cycloheximide ≥90.0%
Cycloheximide ≥90.0%

Glass bottle, 10 g

Numéro CAS:  66-81-9
Prix
550,00 €
Log in for availability
1 * 10 g
Product Details & Documents

Inhibits DNA and protein synthesis.

Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • CAS Number:66-81-9
  • Formula:C₁₅H₂₃NO₄
  • MDL Number:MFCD00082346
  • Molecular Weight:281.35 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-636-0
  • UN:2811
  • ADR:6.1,I
Specifications

Specification Test Results

  • Assay:
    Min. 90.0 %
  • Melting point:
    107 - 120 °C

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.