Cycloheximide 95%
: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
- :66-81-9
- :C₁₅H₂₃NO₄
- :MFCD00082346
- :281.35 g/mol
- :107…114 °C
- :Ambient
- :200-636-0
- :2811
- :6.1,I
: Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
- :66-81-9
- :C₁₅H₂₃NO₄
- :MFCD00082346
- :281.35 g/mol
- :107…114 °C
- :Ambient
- :200-636-0
- :2811
- :6.1,I
- Appearance:White to light beige Powder
- Infrared spectrum:Conforms
- Melting point:108 to 118 °C
- HPLC:≥94 %
- Specific optical rotation:-25° to -32° (20 °C, 589 nm) (c=1, CHCl3)
Product Family Options
- Pack typePk



