Order Entry
Belgium
ContactUsLinkComponent
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, GPR RECTAPUR®
Health
Irritant
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Health
Irritant
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Acidity:
    Max. 0.0002 meq/g
  • Alkalinity:
    Max. 0.003 meq/g
  • Water:
    Max. 0.1 %

Specification Test Results

Frequently Bought Together