Order Entry
Belgium
ContactUsLinkComponent
N-Methyl-2-pyrrolidone (NMP), BAKER ANALYZED®, J.T.Baker®
Health
Irritant
N-Methyl-2-pyrrolidone (NMP), BAKER ANALYZED®, J.T.Baker®
Catalog # 9345.4000GL
Supplier:  Avantor
CAS Number:  
undefined
Health
Irritant
N-Methyl-2-pyrrolidone (NMP), BAKER ANALYZED®, J.T.Baker®
Catalog # 9345.4000GL
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4 L
  • Seq:
    5162

Specifications

Specification Test Results

  • Assay:
    min. 99.5 %
  • Color (APHA):
    max. 20
  • Free Amines (as CH3NH2):
    max. 0.01 %
  • Water (H2O):
    max. 200 ppm

Specification Test Results

About this item

 :
 :99.13 g/mol
Merck Index:13,06140
Boiling Pt:202 °C (1013 hPa)
Melting Pt:–24 °C
Density:1.033 g/cm³ (20 °C)
Flash Pt:91 °C
EINECS:212-828-1
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:872-50-4
Formula:C₅H₉NO
MDL Number:MFCD00003193