Order Entry
Belgium
ContactUsLinkComponent
 
 
 
N-Methyl-2-pyrrolidone (NMP) 99+% for spectroscopy
 
undefined
 
 
N-Methyl-2-pyrrolidone (NMP) 99+% for spectroscopy

 

  • Appearance:
    Clear Liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 % (K.F.)
  • GC:
    ≥99.0 %
  • UV:
    at 285 nm A: ≤1.0
  • Color scale:
    ≤10 APHA

 

 

Frequently Bought Together