4-Hydroxy-3-methoxycinnamic acid (Ferulic acid), Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:537-98-4
:1135-24-6
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00004400
:194.18 g/mol
:M-Clarity™ quality level = MQ100
EINECS:208-679-7
Frequently Bought Together