Order Entry
Belgium
ContactUsLinkComponent
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) for synthesis, Sigma-Aldrich®
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Melting Point:
    169 - 172 °C
  • Molar Mass:
    194.19 g/mol
  • Synonyms:
    Ferulic acid
  • Hill Formula:
    C10H10O4
  • Chemical Formula:
    4-(OH)-3-(OCH3)C6H3CH=CHCOOH

Specification Test Results

Related Information

Frequently Bought Together