Order Entry
Belgium
ContactUsLinkComponent
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) 99%
Irritant
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) 99%
Certificates
undefined
Irritant
4-Hydroxy-3-methoxycinnamic acid (Ferulic acid) 99%
Certificates

Specification Test Results

  • Appearance:
    Off-white to beige Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    169 to 173 °C
  • HPLC:
    ≥98.5 %
  • Titration with NaOH:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together