Maleic acid for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:110-16-7
:
Formula:C₄H₄O₄
MDL Number:MFCD00063177
:116.07 g/mol
Merck Index:13,05726
Boiling Pt:256 °C (1013 hPa)
Melting Pt:130…131 °C
Density:1.59 g/cm³ (20 °C)
EINECS:203-742-5
UN:3261
ADR:8,III
:M-Clarity™ quality level = MQ200
Flash Pt:127 °C (closed cup)
:GHS05
:GHS07
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Solubility:478.8 g/l (20 °C)
- Melting Point:130 - 135 °C
- Molar Mass:116.07 g/mol
- Bulk Density:750 - 800 kg/m3
- Vapor Pressure:- 0.1 hPa (20 °C)
- Flash Point:100 °C
- Density:1.59 g/cm3 (20 °C)
- pH:1.3 (100 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot Numbermsds_800380
SDS
ProductPage_800380 ProductPage_800380
Product specification
Frequently Bought Together