Order Entry
Belgium
ContactUsLinkComponent
 
 
Homovanillic acid fluorimetric reagent, Sigma-Aldrich®
 
undefined
 
Homovanillic acid fluorimetric reagent, Sigma-Aldrich®

 

  • Appearance (Colour):
    White to Off White
  • Appearance (Form):
    Powder
  • Solubility (Colour):
    Colourless to Light Yellow
  • Solubility (Turbidity), 50 mg/ml, EtOH:
    Clear
  • Infrared spectrum:
    Conforms to Structure
  • Titration with NaOH:
    ≥98%
  • Purity (TLC):
    ≥99%
  • Recommended Retest Period:
    4 years

 

Frequently Bought Together