1-Naphthylacetic acid 96% may cont. up to 4% 2-isomer
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:86-87-3
:
Formula:C₁₂H₁₀O₂
MDL Number:MFCD00004046
:186.21 g/mol
Merck Index:14,6371
Boiling Pt:352 °C (1013 hPa)
Melting Pt:126…133 °C
Density:∼1.263 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-705-8
:GHS05
:GHS07
Related Information
Frequently Bought Together