Order Entry
Belgium
ContactUsLinkComponent
Petroleum standard, regular unleaded gasoline 10000 µg/ml in methanol, with certified content of BTEX, Dr. Ehrenstorfer
Petroleum standard, regular unleaded gasoline 10000 µg/ml in methanol, with certified content of BTEX, Dr. Ehrenstorfer
Catalog # EHERYA03001700ME
Supplier:  EHRENSTORFER
CAS Number:  
Petroleum standard, regular unleaded gasoline 10000 µg/ml in methanol, with certified content of BTEX, Dr. Ehrenstorfer
Catalog # EHERYA03001700ME
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Concentration:
    10000 µg/ml in methanol, with certified content of BTEX
  • Pk:
    1 mL
  • Standards:
    Reference standard
  • Description:
    Regular unleaded gasoline

Specifications

About this item

Certifications: ISO/IEC 17025 certified.