Order Entry
Belgium
ContactUsLinkComponent
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
Flammable
Irritant
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Irritant
tert-Butyl methyl ether ≥99%, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay:
    Min. 99 %
  • Appearance:
    Clear colourless liquid
  • IR Spectrum:
    Passes test
  • Water:
    Max. 1 %

Specification Test Results

Frequently Bought Together