Order Entry
Belgium
ContactUsLinkComponent
 
Flammable
Irritant
tert-Butyl methyl ether, Uvasol® for spectroscopy, Supelco®
Catalog # 1.01984.1000
Supplier:  Merck
CAS Number:  
undefined
Flammable
Irritant
tert-Butyl methyl ether, Uvasol® for spectroscopy, Supelco®
Catalog # 1.01984.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    42 g/L (20 °C)
  • Melting Point:
    -108.6 °C
  • Molar Mass:
    88.15 g/mol
  • Boiling Point:
    55.3 °C (1013 hPa)
  • Vapor Pressure:
    268 hPa (20 °C)
  • Flash Point:
    -28 °C
  • Explosion Limit:
    1.5 - 8.5%(V)
  • Density:
    0.74 g/cm³ (20 °C)
  • pH:
    (H₂O) not applicable
  • Ignition Point:
    460 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable liquid

Specification Test Results

About this item

 :
 :88.15 g/mol
Merck Index:13,06059
Boiling Pt:55.2 °C (1013 hPa)
Melting Pt:–108.6 °C
Density:0.7405 g/cm³ (20 °C)
Flash Pt:–28 °C
EINECS:216-653-1
UN:2398
ADR:3,II
 :M-Clarity™ quality level = MQ100
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812