Order Entry
Belgium
ContactUsLinkComponent
tert-Butyl methyl ether ≥99.7%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Irritant
Flammable
tert-Butyl methyl ether ≥99.7%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
Certificates
undefined
Irritant
Flammable
tert-Butyl methyl ether ≥99.7%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™

Specification Test Results

  • Assay (GC):
    Min. 99.7 %
  • Non-volatile matter:
    Max. 0.0005 %
  • Water (Karl Fischer):
    Max. 0.05 %
  • Free acid (as CH₃COOH):
    Max. 0.002 %
  • Peroxides (as H₂O₂):
    Max. 0.0005 %
  • Suitability for residue analysis:
    Max. 5 ng/l
  • Vapor Density:
    3.1 (vs air)
  • Determination of P/N:
    ≤5 ng/L
  • Refractive Index:
    n20/D 1.369 (lit.)
  • Autoignition Temperature:
    705 °F

Specification Test Results

Related Information

Frequently Bought Together