Order Entry
Belgium
ContactUsLinkComponent
tert-Butyl methyl ether ≥99.0%, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Flammable
Irritant
tert-Butyl methyl ether ≥99.0%, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Catalog # 9043-02
Supplier:  Avantor
CAS Number:  
undefined
Flammable
Irritant
tert-Butyl methyl ether ≥99.0%, ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Catalog # 9043-02
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • For Laboratory,Research,or Manufacturing Use:
    = 10
  • Trace Organic Residues - FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):
    ≤10
  • ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):
    ≤10
  • Assay ((CH₃)₃COCH₃) (by GC, corrected for water):
    ≥99.0 %
  • Residue after Evaporation:
    ≤2.0 ppm
  • Peroxide (as H₂O₂):
    ≤1 ppm
  • Water (H₂O)(by coulometry):
    ≤0.05 %

Specification Test Results

About this item

 :
 :88.15 g/mol
Merck Index:13,06059
Boiling Pt:55.2 °C (1013 hPa)
Melting Pt:–108.6 °C
Density:0.7405 g/cm³ (20 °C)
Flash Pt:–28 °C
EINECS:216-653-1
UN:2398
ADR:3,II
 :Packaged under inert gas.
 :GHS02
 :GHS07
Storage Temperature:Ambient
CAS Number:1634-04-4
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812