


Tert-Butyle methyle oxyde ≥99.0% (par GC, corrigé de la teneur en eau), BAKER ANALYZED® HPLC pour HPLC/UHPLC, pour spectrophotométrie, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1634-04-4
:
Formula:(CH₃)₃COCH₃
MDL Number:MFCD00008812
:88.15 g/mol
Merck Index:13,06059
Boiling Pt:55,2 °C (1013 hPa)
Melting Pt:–108,6 °C
Density:0,7405 g/cm³ (20 °C)
Flash Pt:–28 °C
Storage Temperature:Température ambiante
EINECS:216-653-1
UN:2398
ADR:3,II
:GHS02
:GHS07
:GHS08
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:= 99.0 %
- Assay (C₅H₁₂O) (by GC, corrected for water):≥ 99.0 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:≤ 0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:≤ 0.02
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:≤ 0.1
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤ 215
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤ 0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤ 1.0 ppb
- Peroxide (as H₂O₂):≤ 1 ppm
- Residue after Evaporation:≤ 3.0 ppm
- Water (by KF, coulometric):≤ 0.05 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_BAKR9042-03
Product specification
VWR J.T.Baker® High Purity solvents and reagents
Product Literature
Frequently Bought Together