Order Entry
Belgium
ContactUsLinkComponent
1-Heptanesulphonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC
1-Heptanesulphonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
Certificates
undefined
1-Heptanesulphonic acid sodium salt ≥99%, HiPerSolv CHROMANORM® for HPLC
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (calculated on dried substance):
    ≥ 99,0 %
  • Appearance:
    White powder
  • IR Spectrum:
    Passes test
  • Water:
    ≤ 2,0 %
  • Absorbance (200 nm) (0.25 mol/l):
    ≤ 0,1
  • Absorbance (210 nm) (0.25 mol/l):
    ≤ 0,05
  • Absorbance (220 nm) (0.25 mol/l):
    ≤ 0,04
  • Absorbance (230 nm) (0.25 mol/l):
    ≤ 0,03
  • Absorbance (240 nm) (0.25 mol/l):
    ≤ 0,01
  • Absorbance (250 nm) (0.25 mol/l):
    ≤ 0,01
  • Absorbance (260 nm) (0.25 mol/l):
    ≤ 0,01

Specification Test Results

Frequently Bought Together