
1-Heptanesulphonic acid sodium salt 98%
:22767-50-6
:
:H₃C(CH₂)₅CH₂SO₃Na
:MFCD00007543
:202.25 g/mol
:>300 °C
:245-210-5
:GHS07
:Ambient
- Appearance:White Powder or flakes
- Infrared spectrum:Conforms
- Loss on drying:≤9 % (100°C, 3 h) (vacuum)
- Solubility:(10 % in water) Clear colorless
- Titration after Ion exchange:≥97.5 % (on dry substance)
Frequently Bought Together