Order Entry
Belgium
ContactUsLinkComponent
Standard mixture of n-alkanes for system performance test
Standard mixture of n-alkanes for system performance test
Catalog # REST31633
Supplier:  Restek
CAS Number:  
Standard mixture of n-alkanes for system performance test
Catalog # REST31633
Supplier:  Restek
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Description:
    Standard mixture of n-alkanes for system performance test, 16 components

Specifications

About this item

These are reference mixtures for ISO/DIS 9377 (German H-53), a gas chromatography–flame ionization detection (GC-FID) method.

  • For GC analysis of total petroleum hydrocarbons (TPH) in water
  • Environmentally safer than previous methods
  • Calibration standard available as diesel no.2/motor oil and diesel no.2/mineral oil

Concentration: 50 μg/ml each in cyclohexane.

Certifications: ISO/DIS 9377 Water Quality Testing (German H-53)