Order Entry
Belgium
ContactUsLinkComponent
 
Sodium-1-decanesulphonate
Catalog # MOLEM15618664
Supplier:  Molekula
CAS Number:  
Sodium-1-decanesulphonate
Catalog # MOLEM15618664
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :244.33 g/mol
Melting Pt:> 300 °C
Storage Temperature:Ambient
EINECS:236-525-9
CAS Number:13419-61-9
Formula:C₁₀H₂₁NaO₃S
MDL Number:MFCD00007526