Sodium-1-decanesulphonate
Catalog # MOLEM15560173
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:244.33 g/mol
Melting Pt:> 300 °C
Storage Temperature:Ambient
EINECS:236-525-9
CAS Number:13419-61-9
Formula:C₁₀H₂₁NaO₃S
MDL Number:MFCD00007526