Order Entry
Belgium
ContactUsLinkComponent
Sodium-1-decanesulphonate for synthesis, Sigma-Aldrich®
Sodium-1-decanesulphonate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Sodium-1-decanesulphonate for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Melting Point:
    300 °C
  • Molar Mass:
    244.33 g/mol

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together