Sodium-1-decanesulphonate, (max. 1,5% H₂O) 99% (dry weight)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13419-61-9
:
Formula:C₁₀H₂₁NaO₃S
MDL Number:MFCD00007526
:244.33 g/mol
Melting Pt:>300 °C
Storage Temperature:Ambient
EINECS:236-525-9
Related Information
Frequently Bought Together