Orders LinkContactUsLinkComponent
VWR® Basic, Visitor Coats, SPP, with Pockets
VWR® Basic, Visitor Coats, SPP, with Pockets
Supplier:   VWR Collection
VWR® Basic, Visitor Coats, SPP, with Pockets
Supplier:  VWR Collection
Certificates
VWR® Basic, Visitor Coats, SPP, with Pockets
6 Options Available Below
Product Details & Documents
These disposable visitor coats, with collar, are made from non woven, SPP fabric.
  • Press stud closure
  • Elastic wrists
  • One breast pocket or one breast and one side pocket
  • Free from natural rubber latex
Weight: 30 or 40 g/m²
Packaging: 1 piece per bag, 50 bags per case.
Specifications for Product Family
Specifications
  • Catalog No:
  • 113-8972
  • 113-8973
  • 113-8974
  • 113-8979
  • 113-8980
  • 113-8982
  • Model:
  • With 1 pocket
  • With 1 pocket
  • With 1 pocket
  • With 2 pockets
  • With 2 pockets
  • With 2 pockets
  • Closure type:
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Press studs / snaps
  • Pockets:
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • With pockets
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Garment application:
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • General Purpose|Laboratory
  • Weight:
  • 30 g/m²
  • 30 g/m²
  • 30 g/m²
  • 40 g/m²
  • 40 g/m²
  • 40 g/m²
Specifications
Product Family Options
With 1 pocket, 30 g/m²
Product Information
  • Colour
    Clothing Size
    Length
    Availability
    Price
  • White
    L
    104 cm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 179,00 €
    179,00 €
    1 * 50 ST
       179,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-8972
    Model
    With 1 pocket
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • White
    XL
    108 cm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 179,00 €
    179,00 €
    1 * 50 ST
       179,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-8973
    Model
    With 1 pocket
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
  • White
    XXL
    112 cm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 179,00 €
    179,00 €
    1 * 50 ST
       179,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-8974
    Model
    With 1 pocket
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    30 g/m²
With 2 pockets, 40 g/m²
Product Information
  • Colour
    Clothing Size
    Length
    Availability
    Price
  • White
    L
    104 cm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 197,00 €
    197,00 €
    1 * 50 ST
       197,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-8979
    Model
    With 2 pockets
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • White
    XL
    108 cm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 197,00 €
    197,00 €
    1 * 50 ST
       197,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-8980
    Model
    With 2 pockets
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²
  • White
    XXL
    112 cm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 197,00 €
    197,00 €
    1 * 50 ST
       197,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    113-8982
    Model
    With 2 pockets
    Closure type
    Press studs / snaps
    Pockets
    With pockets
    Gender
    Unisex
    Garment application
    General Purpose|Laboratory
    Weight
    40 g/m²