
1-Octadecanethiol 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2885-00-9
:
Formula:C₁₈H₃₈S
MDL Number:MFCD00004886
:286.57 g/mol
Boiling Pt:366 °C (760 mmHg)
Melting Pt:25…35 °C
Density:0,8475 g/cm³ (20 °C)
Flash Pt:185 °C
Storage Temperature:Ambient
EINECS:220-744-1
UN:3335
ADR:9,III
:GHS07
Specification Test Results
- Appearance:White Crystalline solid
- Infrared spectrum:Conforms
- Melting point:25 to 35 °C
- GC:≥95 %
Specification Test Results
Frequently Bought Together