Order Entry
Belgium
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) ≥94.0%, TECHNICAL
Flammable
Irritant
Corrosive
Sodium dodecyl sulphate (SDS) ≥94.0%, TECHNICAL
Catalog # 83886.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
Corrosive
Sodium dodecyl sulphate (SDS) ≥94.0%, TECHNICAL
Catalog # 83886.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    Min. 94,0 %
  • Sodium chloride:
    Max. 1.0 %
  • Sodium sulphate:
    Max. 2.5 %

Specification Test Results

About this item

 :
 :288.38 g/mol
Merck Index:14,08636
Melting Pt:204…207 °C
Density:1.1 g/cm³ (20 °C)
Flash Pt:100 °C (closed cup)
Storage Temperature:Ambient
EINECS:205-788-1
UN:1325
ADR:4.1,III
REACH:01-2119489461-32
 :GHS02
 :GHS07
 :GHS05
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175