
Ethylene glycol
Supplier: Roth Carl
Certificates
About this item
CAS Number:107-21-1
:
Formula:HOCH₂CH₂OH
MDL Number:MFCD00002885
:62.07 g/mol
Merck Index:13,03832
Boiling Pt:196…198 °C (1013 hPa)
Melting Pt:–13 °C
Density:1.115 g/cm³ (20 °C)
Flash Pt:116 °C
Storage Temperature:Refrigerator
EINECS:203-473-3
REACH:01-2119456816-28
:GHS07
Frequently Bought Together