Order Entry
Belgium
ContactUsLinkComponent
Ethanol ≥96% (v/v) Reag. Ph. Eur., Supelco®
Flammable
Irritant
Ethanol ≥96% (v/v) Reag. Ph. Eur., Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates
Flammable
Irritant
Ethanol ≥96% (v/v) Reag. Ph. Eur., Supelco®
Supplier:  Merck
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Specification Test Results

  • Grade:
    Reag. Ph Eur
  • Hill Formula:
    C₂H₆O
  • Chemical formula:
    C₂H₅OH
  • Molar Mass:
    46.07 g/mol
  • Boiling point:
    78 °C (1013 hPa)
  • Density:
    0.805 - 0.812 g/cm³ (20 °C)
  • Explosion limit:
    3.5 - 15 %(V)
  • Flash point:
    17 °C
  • Ignition temperature:
    425 °C
  • Melting Point:
    -117 °C
  • pH value:
    7.0 (10 g/l, H₂O, 20 °C)
  • Vapor pressure:
    59 hPa (20 °C)
  • Solubility:
    (20 °C) soluble
  • LD 50 oral:
    LD50 Rat 6200 mg/kg
  • Storage class:
    3 Flammable liquids
  • WGK:
    WGK 1 slightly water endangering
  • R Phrase:
    R 11
  • S Phrase:
    S 7-16

Specification Test Results

Related Information

Frequently Bought Together