Order Entry
Belgium
ContactUsLinkComponent
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Flammable
Irritant
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates
undefined
Flammable
Irritant
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier:  VWR Chemicals
   Sustainable Options Available
Certificates

Specifications

  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion

Specifications

Specification Test Results

  • Assay (on anhydrous substance):
    Min. 99.8 %
  • Assay (V/V):
    95.1 - 96.9 %
  • IR Spectrum:
    Passes test
  • UV Spectrum:
    Passes test
  • Acidity:
    Max. 0.0002 meq/g
  • Alkalinity:
    Max. 0.0001 meq/g
  • Boiling point:
    78.0 - 78.5 °C
  • Colouration:
    Max. 10 APHA
  • Substances coloured by H2SO4:
    Max. 20 APHA
  • Aldehydes (as CH3CHO):
    Max. 5 ppm
  • Aldehydes + ketones (as CH3CHO):
    Max. 10 ppm
  • Benzene:
    Max. 5 ppm
  • Evaporation residue:
    Max. 5 ppm
  • Formaldehyde:
    Max. 1 ppm
  • Furaldehyde:
    Max. 1 ppm
  • Ketones (as CH3COCH3):
    Max. 10 ppm
  • Methanol:
    Max. 0.02 %
  • Propan-2-ol:
    Max. 100 ppm
  • Substances reducing KMnO4 (as O):
    Max. 4 ppm
  • Total heavy alcohols:
    Max. 100 ppm
  • Water:
    5.2 - 8.2 %
  • Al (Aluminium):
    Max. 0.1 ppm
  • Ba (Barium):
    Max. 0.02 ppm
  • Ca (Calcium):
    Max. 0.2 ppm
  • Cd (Cadmium):
    Max. 0.02 ppm
  • Co (Cobalt):
    Max. 0.01 ppm
  • Cr (Chromium):
    Max. 0.01 ppm
  • Cu (Copper):
    Max. 0.1 ppm
  • Fe (Iron):
    Max. 0.05 ppm
  • K (Potassium):
    Max. 0.1 ppm
  • Mg (Magnesium):
    Max. 0.05 ppm
  • Mn (Manganese):
    Max. 0.01 ppm
  • Na (Sodium):
    Max. 1 ppm
  • Ni (Nickel):
    Max. 0.02 ppm
  • Pb (Lead):
    Max. 0.05 ppm
  • Sn (Tin):
    Max. 0.05 ppm
  • Sr (Strontium):
    Max. 0.02 ppm
  • Zn (Zinc):
    Max. 0.1 ppm

Specification Test Results

Frequently Bought Together