

Ethanol (untaxed) ~96%, Puriss. p.a. ACS, without additive prima fine spirit, F15 (not denatured), Riedel-de Haën™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:64-17-5
:
Formula:C₂H₅OH
MDL Number:MFCD00003568
:46.07 g/mol
Boiling Pt:78…79 °C (1.013 hPa)
Melting Pt:–115 °C
Density:0.805…0.812 g/cm3 (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-578-6
UN:1170
ADR:3,II
:GHS07
:GHS02
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
Specification Test Results
- Appearance of the substance:complying
- Assay (GC):Min. 99.8 %
- Refractive index (n 20/D):1.363 - 1.365
- Non-volatile matter:Max. 0.001 %
- Water (Karl Fischer):5.0 - 7.0 %
- Titrable Acid:Max. 0.0005 meq/g
- Titrable Base:Max. 0.0002 meq/g
- Aluminium (Al):Max. 0.5 ppm
- Barium (Ba):Max. 0.1 ppm
- Bismuth (Bi):Max. 0.1 ppm
- Calcium (Ca):Max. 0.5 ppm
- Cadmium (Cd):Max. 0.05 ppm
- Cobalt (Co):Max. 0.02 ppm
- Chromium (Cr):Max. 0.02 ppm
- KMnO₄ reducing matter:complying
- Acetone (GC):Max. 10 ppm
- Aldehydes (as CH₃CHO):Max. 10 ppm
- Formaldehyde:Max. 10 ppm
- Methanol (GC):Max. 0.1 %
- 2-propanol (GC):Max. 30 ppm
- Reaction against H₂SO₄:complying
- Mixable with H₂O:complying
- APHA:Max. 10
- Vapor Pressure:44.6 mmHg ( 20 °C)
- Vapor Density:1.59 (vs air)
Specification Test Results
Frequently Bought Together