

2-Propanol ≥99.0%, meets analytical specification of USP, Riedel-de Haën™
Catalog # I9030-500ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Vapor Density:2.1 (vs air)
- Assay (GC):Min. 99.0 %
- Density (D 25/25):0.783 - 0.787
- Refractive index (n 20/D):1.376 - 1.378
- Water (Karl Fischer):Max. 0.5 %
- Non-volatile matter:Max. 0.005 %
- Acidity:complying
- Volatile impurities (GC):complying
- Acetone (GC):Max. 0.1 %
- 2-Butanol (GC):Max. 0.1 %
- Diethyl ether (GC):Max. 0.1 %
- Diisopropyl alcohol:Max. 0.1 %
- 1-propanol (GC):Max. 0.1 %
- Impurities (GC) total:Max. 1.0 %
- Residual solvents:complying
Specification Test Results
About this item
:
:60.1 g/mol
Merck Index:13,05228
Boiling Pt:77…83 °C (1.013 hPa)
Melting Pt:–90 °C
Density:0.784…0.786 g/cm³ (20 °C)
Flash Pt:12 °C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
:GHS07
:GHS02
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674