Order Entry
Belgium
ContactUsLinkComponent
1-Butanol ≥99.9%
Flammable
Corrosive
Irritant
1-Butanol ≥99.9%
Certificates
undefined
Flammable
Corrosive
Irritant
1-Butanol ≥99.9%
Certificates

Specification Test Results

  • Identity (IR):
    complying
  • Assay (GC):
    Min. 99.80 %
  • Density (D 20/4):
    0.809 - 0.811
  • Non-volatile matter:
    Max. 20 mg/l
  • Water (Karl Fischer):
    Max. 0.1 %
  • Free acid (as CH₃COOH):
    Max. 0.005 %
  • Iron (Fe):
    Max. 0.3 ppm
  • Di-n-butyl ether (GC):
    Max. 0.100 %
  • Isobutyl alcohol (GC):
    Max. 0.100 %
  • n-Butyraldehyde (GC):
    Max. 0.020 %
  • Carbonyl compounds (as CO):
    Max. 0.050 %
  • APHA:
    Max. 10
  • Autoignition Temperature:
    649 °F
  • Refractive Index:
    n20/D 1.399 (lit.)

Specification Test Results

Related Information

Frequently Bought Together