Order Entry
Belgium
ContactUsLinkComponent
Acetic anhydride, BAKER ANALYZED® ACS, J.T.Baker®
Flammable
Corrosive
Irritant
Acetic anhydride, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates
undefined
Flammable
Corrosive
Irritant
Acetic anhydride, BAKER ANALYZED® ACS, J.T.Baker®
Supplier:  Avantor
Certificates

Specification Test Results

  • Assay:
    min. 97.0 %
  • Phosphate (PO4):
    max. 0.001 %
  • Residue after Evaporation:
    max. 0.003 %
  • Substances Reducing KMnO4:
    passes test
  • Trace Impurities (in ppm) Chloride (Cl):
    max. 5
  • Trace Impurities (in ppm) Iron (Fe):
    max. 5
  • Trace Impurities (in ppm) Heavy Metals (as Pb):
    max. 2
  • Trace Impurities (in ppm) Sulfate (SO4):
    max. 5

Specification Test Results

Related Information

Frequently Bought Together