Diisopropyl ether, EMSURE® Reag. Ph. Eur., ACS for analysis, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Isopropyl ether
CAS Number:108-20-3
:
Formula:(CH₃)₂CHOCH(CH₃)₂
MDL Number:MFCD00008880
:102.18 g/mol
Merck Index:13,05232
Boiling Pt:68.5 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.726 g/cm³ (20 °C)
EINECS:203-560-6
UN:1159
ADR:3,II
:M-Clarity™ quality level = MQ300
Flash Pt:–28 °C
:GHS02
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:12 g/l (20 °C)
- Melting Point:-86 °C
- Molar Mass:102.18 g/mol
- Boiling Point:67 - 70 °C (1013 hPa)
- Vapor Pressure:175 hPa (20 °C)
- Flash Point:-28 °C
- Refractive Index:1.3679 (20 °C, 589 nm)
- Explosion Limit:1.0 - 21 %(V)
- Density:0.72 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral
- Ignition Point:405 °C
- Evaporation number:1.6
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.00867.1000_TDS_EN ProductPage_100867
Product specification
msds_100867
SDS
Frequently Bought Together