Bis-trispropane
Marketsource
Supplier: Roth Carl
Jump to:
About this item
Synonyms: 1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]
- CAS Number:64431-96-5
- Formula:C₁₁H₂₆N₂O₆
- MDL Number:MFCD00004689
- Molecular Weight:282.34 g/mol
- EINECS:264-899-3
2 Options Available Below
Product Details & Documents
Synonyms: 1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]
- CAS Number:64431-96-5
- Formula:C₁₁H₂₆N₂O₆
- MDL Number:MFCD00004689
- Molecular Weight:282.34 g/mol
- EINECS:264-899-3
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:MarketsourceMarketsource
- Specifications:MarketsourceMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



