Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Bis-trispropane

Glass bottle for solids, 100 g
Catalog # MOLEM12065389
Marketsource
Supplier:   Molekula
Bis-trispropane
Catalog # MOLEM12065389
Supplier:  Molekula
Marketsource
CAS Number:  64431-96-5
Bis-trispropane

Glass bottle for solids, 100 g

CAS Number:  64431-96-5
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  1,3-Bis(tris(hydroxymethyl)methylamino)propane, 1,3-Bis[tris(hydroxymethyl)amino]propane, 2,2'-(Propane-1,3-diylbis(azanediyl))bis(2-(hydroxymethyl)propane-1,3-diol), BIS-TRIS propane, 2,2'-(Propane-1,3-diyldiimino)bis[2-(hydroxymethyl)propane-1,3-diol]
  • CAS Number:64431-96-5
  • Formula:C₁₁H₂₆N₂O₆
  • MDL Number:MFCD00004689
  • Molecular Weight:282.34 g/mol
  • EINECS:264-899-3
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.