Order Entry
Belgium
ContactUsLinkComponent
 
Diethanolamine, EMSURE® for analysis, Supelco®
Catalog # 1.16205.1000
Supplier:  Merck
CAS Number:  
Diethanolamine, EMSURE® for analysis, Supelco®
Catalog # 1.16205.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    28 °C
  • Molar Mass:
    105.14 g/mol
  • Boiling Point:
    269 - 270 °C (1013 hPa)
  • Vapor Pressure:
     - 0.01 hPa (25 °C)
  • Flash Point:
    176 °C
  • Refractive Index:
    1.4753 (30 °C, 589 nm)
  • Explosion Limit:
    2.1 - 10.6 %(V)
  • Density:
    1.09 g/cm3 (30 °C) liquid
  • pH:
    11 (53 g/l, H2O, 20 °C)
  • Ignition Point:
    365 - 370 °C

Specification Test Results

About this item

 :
 :105.14 g/mol
Merck Index:13,03134
Boiling Pt:268.8 °C (1013 hPa)
Melting Pt:28 °C
Density:1.1 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ300
Flash Pt:176 °C
EINECS:203-868-0
REACH:01-2119488930-28
 :GHS05
 :GHS08
 :GHS07
Storage Temperature:Ambient
CAS Number:111-42-2
Formula:C₄H₁₁NO₂
MDL Number:MFCD00002843