


Diethanolamine, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:111-42-2
:
Formula:C₄H₁₁NO₂
MDL Number:MFCD00002843
:105.14 g/mol
Merck Index:13,03134
Boiling Pt:268.8 °C (1013 hPa)
Melting Pt:28 °C
Density:1.1 g/cm³ (20 °C)
Flash Pt:176 °C
Storage Temperature:Ambient
EINECS:203-868-0
REACH:01-2119488930-28
:GHS05
:GHS08
:GHS07
Specification Test Results
- Assay:min. 98.5 %
- Density (g/ml) at 30°C:1.088 - 1.093
- Iron (Fe):max. 0.001 %
- Heavy Metals (as Pb):max. 0.002 %
- Residue after Ignition:max. 0.005 %
- Trace Impurities (in ppm) Chloride (Cl):max. 5
Specification Test Results
Frequently Bought Together